siRNA Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011149.2 | NP_035279.2 | 2 | 314 | ||
GenBank mRNA | AK002357.1 | - | 2 | 261 | ||
AK128977.1 | - | 2 | 264 | |||
AK154073.1 | - | 2 | 253 | |||
AL363449.1 | - | 2 | 355 | |||
BC013061.1 | - | 2 | 251 | 3483267 | ||
M60456.1 | - | 2 | 233 | |||
X58990.1 | - | 2 | 359 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022536, FQ231656, FQ228087, FQ222696, FQ230004, and others.
Gene Ontology Categories:
