siRNA Details
Gene Symbol: Lgals3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011150.2 | NP_035280.1 | 6 | 1470 | ||
GenBank mRNA | AK079915.1 | - | 6 | 1499 | ||
AK137836.1 | - | 6 | 1447 | |||
AK150021.1 | - | 6 | 1499 | |||
AK152182.1 | - | 6 | 1412 | |||
AK153313.1 | - | 6 | 1486 | |||
AK153396.1 | - | 6 | 1488 | |||
AK155031.1 | - | 6 | 1412 | |||
BC090658.1 | - | 6 | 1466 | 30666981 | ||
L16894.1 | - | 6 | 1477 | |||
X67809.1 | - | 6 | 1470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
