siRNA Details
Gene Symbol: Ppm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008910.3 | NP_032936.1 | 5 | 1527 | ||
GenBank mRNA | AK208896.1 | - | 1 | 24 | ||
AK218433.1 | - | 1 | 24 | |||
BC008595.1 | - | 5 | 1134 | 3581897 | ||
D28117.1 | - | 5 | 1203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
