siRNA Details
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008914.2 | NP_032940.1 | 5, 6 | 799 | ||
GenBank mRNA | AB221678.1 | - | 5, 6 | 666 | ||
BC037668.1 | - | 5, 6 | 622 | |||
BC066000.1 | - | 5, 6 | 770 | 5699179 | ||
M81483.1 | - | 5, 6 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
