siRNA Details
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015002.1 | - | 8 | 1110 | ||
AK133393.1 | - | 8 | 1131 | |||
BC046617.1 | - | 8 | 936 | |||
BC141079.1 | - | 8 | 951 | 9055691 | ||
BC145347.1 | - | 7 | 826 | |||
M81475.1 | - | 8 | 1104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
