siRNA Details
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000618.4 | NP_000609.1 | 4 | 1680 | ||
NM_001111283.2 | NP_001104753.1 | 5 | 1729 | |||
NM_001111284.1 | NP_001104754.1 | 4 | 1518 | |||
XM_017019262.1 | XP_016874751.1 | 5 | 1539 | |||
XM_017019263.1 | XP_016874752.1 | 4 | 1482 | |||
GenBank mRNA | AB209184.1 | - | 4 | 1631 | ||
BC152321.1 | - | 4 | 1561 | 40129266 | ||
X57025.1 | - | 4 | 1581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
