siRNA Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008917.3 | NP_032943.2 | 5 | 541 | ||
GenBank mRNA | AF071025.1 | - | 5 | 558 | ||
AF087568.1 | - | 5 | 519 | |||
AF326557.1 | - | 5 | 525 | |||
AF326558.1 | - | 5 | 525 | |||
AK014561.1 | - | 5 | 523 | |||
AK018046.1 | - | 1 | 1019 | |||
AK148314.1 | - | 5 | 516 | |||
AK151055.1 | - | 5 | 516 | |||
AK151107.1 | - | 5 | 527 | |||
AK152474.1 | - | 5 | 525 | |||
AK152940.1 | - | 5 | 527 | |||
AK153316.1 | - | 5 | 522 | |||
AK169579.1 | - | 6 | 616 | |||
AK169658.1 | - | 6 | 622 | |||
AK171668.1 | - | 5 | 523 | |||
AK216167.1 | - | 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
