siRNA Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008917.3 | NP_032943.2 | 5 | 552 | ||
GenBank mRNA | AF071025.1 | - | 5 | 569 | ||
AF087568.1 | - | 5 | 530 | |||
AF326557.1 | - | 5 | 536 | |||
AF326558.1 | - | 5 | 536 | |||
AK014561.1 | - | 5 | 534 | |||
AK018046.1 | - | 1 | 1030 | |||
AK148314.1 | - | 5 | 527 | |||
AK151055.1 | - | 5 | 527 | |||
AK151107.1 | - | 5 | 538 | |||
AK152474.1 | - | 5 | 536 | |||
AK152940.1 | - | 5 | 538 | |||
AK153316.1 | - | 5 | 533 | |||
AK169579.1 | - | 6 | 627 | |||
AK169658.1 | - | 6 | 633 | |||
AK171668.1 | - | 5 | 534 | |||
AK216167.1 | - | 3 | 204 |
