siRNA Details
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008922.2 | NP_032948.1 | 3 | 384 | ||
GenBank mRNA | AK076095.1 | - | 3 | 366 | ||
AK077760.1 | - | 3 | 371 | |||
AK153970.1 | - | 3 | 345 | |||
AK168246.1 | - | 3 | 377 | |||
BC019500.1 | - | 3 | 345 | 3602929 | ||
D13545.1 | - | 3 | 348 | |||
D17385.1 | - | 3 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
