siRNA Details
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008922.2 | NP_032948.1 | 5 | 541 | ||
GenBank mRNA | AK076095.1 | - | 5 | 523 | ||
AK077760.1 | - | 5 | 528 | |||
AK153970.1 | - | 5 | 502 | |||
AK168246.1 | - | 5 | 534 | |||
BC019500.1 | - | 5 | 502 | 3602929 | ||
D13545.1 | - | 5 | 505 | |||
D17385.1 | - | 5 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
