siRNA Details
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011159.2 | NP_035289.2 | 15 | 1588 | ||
GenBank mRNA | AB007544.1 | - | 15 | 1565 | ||
AB011543.1 | - | 15 | 1577 | |||
AK084827.1 | - | 15 | 1589 | |||
D87521.1 | - | 15 | 1565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
