siRNA Details
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011159.2 | NP_035289.2 | 24, 25 | 2791 | ||
GenBank mRNA | AB007544.1 | - | 24, 25 | 2768 | ||
AB011543.1 | - | 24, 25 | 2780 | |||
D87521.1 | - | 24, 25 | 2768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
