siRNA Details
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202445.1 | NP_001189374.1 | 3, 4 | 946 | ||
NM_001202446.1 | NP_001189375.1 | 3, 4 | 871 | |||
NM_011162.5 | NP_035292.2 | 3, 4 | 654 | |||
GenBank mRNA | AB221603.1 | - | 3, 4 | 517 | ||
AF003115.1 | - | 3, 4 | 654 | |||
AF054611.1 | - | 3, 4 | 654 | |||
AF109768.1 | - | 3, 4 | 551 | |||
AF109769.1 | - | 3, 4 | 614 | |||
AF109770.1 | - | 3, 4 | 669 | |||
AF109771.1 | - | 3, 4 | 515 | |||
AF332075.1 | - | 3, 4 | 517 | |||
AF332076.1 | - | 3, 4 | 517 | |||
AK147033.1 | - | 3, 4 | 873 | |||
BC072578.1 | - | 3, 4 | 838 | 6402485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
