siRNA Details
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011163.4 | NP_035293.1 | 6 | 751 | ||
GenBank mRNA | AK028602.1 | - | 6 | 751 | ||
BC016422.1 | - | 6 | 714 | 3964935 | ||
M65029.1 | - | 6 | 703 | |||
M93567.1 | - | 6 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
