siRNA Details
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163530.1 | NP_001157002.1 | 2, 3 | 260 | ||
NM_011164.2 | NP_035294.2 | 2, 3 | 263 | |||
GenBank mRNA | AK017520.1 | - | 2, 3 | 256 | ||
AK017521.1 | - | 2, 3 | 257 | |||
AK017547.1 | - | 2, 3 | 261 | |||
AK017562.1 | - | 2, 3 | 248 | |||
AK017579.1 | - | 2, 3 | 257 | |||
AK017584.1 | - | 2, 3 | 254 | |||
AK077493.1 | - | 2, 3 | 260 | |||
AK133645.1 | - | 2, 3 | 254 | |||
AK133736.1 | - | 2, 3 | 257 | |||
AK133752.1 | - | 2, 3 | 257 | |||
AK161553.1 | - | 2, 3 | 253 | |||
AK161557.1 | - | 2, 3 | 257 | |||
AK161565.1 | - | 2, 3 | 257 | |||
AK161594.1 | - | 2, 3 | 254 | |||
AK161619.1 | - | 2, 3 | 261 | |||
BC061141.1 | - | 2, 3 | 256 | 30250937 | ||
X02892.1 | - | 2, 3 | 255 | |||
X04418.1 | - | 2, 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
