siRNA Details
Gene Symbol: Klk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164696.1 | NP_001158168.1 | 5 | 577 | ||
NM_001164697.1 | NP_001158169.1 | 5 | 573 | |||
NM_001164698.1 | NP_001158170.1 | 5 | 474 | |||
NM_011177.2 | NP_035307.2 | 4 | 585 | |||
GenBank mRNA | AB008928.1 | - | 5 | 563 | ||
AB015206.1 | - | 5 | 561 | |||
AK138218.1 | - | 5 | 574 | |||
BC031119.1 | - | 4 | 422 | 4954976 | ||
EL956218.1 | - | 5 | 449 | |||
Y18723.1 | - | 4 | 585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
