siRNA Details
Gene Symbol: Prtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011178.2 | NP_035308.2 | 5 | 924 | ||
GenBank mRNA | FM210470.1 | - | 3 | 738 | ||
U43525.1 | - | 5 | 912 | |||
U97073.1 | - | 5 | 924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
