siRNA Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008943.2 | NP_032969.1 | 7 | 1075 | ||
GenBank mRNA | AK134232.1 | - | 7 | 758 | ||
AK134615.1 | - | 7 | 797 | |||
AK148101.1 | - | 7 | 796 | |||
AK165984.1 | - | 7 | 796 | |||
AK169963.1 | - | 3 | 202 | |||
BC014744.1 | - | 7 | 826 | 4511140 | ||
BC030409.1 | - | 7 | 750 | 5133302 | ||
BC071233.1 | - | 7 | 814 | 3673609 | ||
L42177.1 | - | 7 | 771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
