siRNA Details
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011184.4 | NP_035314.3 | 4 | 385 | ||
GenBank mRNA | AF055983.1 | - | 4 | 303 | ||
AF060089.1 | - | 4 | 271 | |||
AI303320.1 | - | 4 | 384 | |||
AK002877.1 | - | 5 | 316 | |||
AK165424.1 | - | 4 | 338 | |||
AK168495.1 | - | 4 | 290 | |||
AK169577.1 | - | 4 | 312 | |||
BC091743.1 | - | 4 | 275 | 5030475 | ||
BY159585.1 | - | 4 | 386 |
