siRNA Details
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011184.4 | NP_035314.3 | 4 | 378 | ||
GenBank mRNA | AF055983.1 | - | 4 | 296 | ||
AF060089.1 | - | 4 | 264 | |||
AI303320.1 | - | 4 | 377 | |||
AK002877.1 | - | 5 | 309 | |||
AK165424.1 | - | 4 | 331 | |||
AK168495.1 | - | 4 | 283 | |||
AK169577.1 | - | 4 | 305 | |||
BC091743.1 | - | 4 | 268 | 5030475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
