siRNA Details
Gene Symbol: Bpifa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008953.2 | NP_032979.1 | 3, 4 | 305 | ||
GenBank mRNA | AK009032.1 | - | 3, 4 | 317 | ||
AK009654.1 | - | 3, 4 | 317 | |||
AK010181.1 | - | 3, 4 | 317 | |||
BC010288.1 | - | 3, 4 | 305 | 2616413 | ||
BC037124.1 | - | 3, 4 | 326 | |||
BC053336.1 | - | 3, 4 | 305 | 4190594 | ||
X01697.1 | - | 2, 3 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
