siRNA Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI021584.1 | - | 2 | 268 | ||
AK075604.1 | - | 5 | 1123 | |||
BC003793.1 | - | 5 | 1093 | 3601675 | ||
U28016.1 | - | 5 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
