siRNA Details
Gene Symbol: Ptgdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008962.4 | NP_032988.3 | 2 | 1113 | ||
GenBank mRNA | AK032488.1 | - | 2 | 1114 | ||
BC132297.1 | - | 2 | 1092 | 40130574 |
Gene Symbol: Gm16976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138521.1 | - | 3 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
