siRNA Details
Gene Symbol: Ptger3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011196.2 | NP_035326.2 | 1 | 466 | ||
GenBank mRNA | BC058742.1 | - | 1 | 466 | 6309451 | |
D10204.1 | - | 1 | 458 | |||
D13321.1 | - | 1 | 458 | |||
D17406.1 | - | 1 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
