siRNA Details
Gene Symbol: Pth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020623.2 | NP_065648.1 | 3 | 240 | ||
GenBank mRNA | AK138677.1 | - | 3 | 238 | ||
BC099456.1 | - | 3 | 225 | 30845904 |

More Information
Other Species Targeted

Gene Ontology Categories:
