siRNA Details
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008972.2 | NP_032998.1 | 5 | 1032 | ||
GenBank mRNA | AK020558.1 | - | 1 | 382 | ||
AK037980.1 | - | 5 | 1044 | |||
AK088005.1 | - | 5 | 1043 | |||
AK143576.1 | - | 5 | 1043 | |||
AK146454.1 | - | 5 | 1040 | |||
AK146651.1 | - | 5 | 1043 | |||
AK146849.1 | - | 5 | 1043 | |||
AK151289.1 | - | 5 | 1042 | |||
AK151511.1 | - | 5 | 1046 | |||
AK151768.1 | - | 5 | 1044 | |||
AK152032.1 | - | 5 | 1042 | |||
AK153178.1 | - | 5 | 1044 | |||
AK153496.1 | - | 5 | 1043 | |||
AK159128.1 | - | 5 | 1043 | |||
AK159170.1 | - | 2 | 3385 | |||
BC083135.1 | - | 5 | 995 | 6506450 | ||
BC083163.1 | - | 1 | 341 | |||
BC085171.1 | - | 5 | 1032 | 30699339 | ||
BC086804.1 | - | 334 | ||||
X56135.1 | - | 5 | 1031 |
Gene Symbol: Gm12504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038173.1 | - | 1 | 2173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
