siRNA Details
Gene Symbol: Tmsb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039392.2 | NP_001034481.1 | 2 | 574 | ||
NM_001190327.1 | NP_001177256.1 | 4 | 585 | |||
NM_025284.4 | NP_079560.1 | 3 | 513 | |||
GenBank mRNA | AK003178.1 | - | 3 | 430 | ||
AK005512.1 | - | 3 | 430 | |||
AK008557.1 | - | 3 | 430 | |||
AK012361.1 | - | 3 | 430 | |||
AK019212.1 | - | 3 | 430 | |||
AK019232.1 | - | 3 | 431 | |||
AK131711.1 | - | 3 | 430 | |||
AK161987.1 | - | 3 | 430 | |||
BC037153.1 | - | 3 | 416 | |||
BC038926.1 | - | 3 | 431 | |||
BC048070.1 | - | 3 | 409 | |||
BC059777.1 | - | 3 | 417 | |||
BC070416.1 | - | 3 | 408 | |||
BC080312.1 | - | 3 | 411 | |||
BC089326.1 | - | 3 | 389 | |||
BC089327.1 | - | 2 | 369 | |||
BC092009.1 | - | 3 | 414 | |||
BC093587.1 | - | 3 | 415 | |||
BC094284.1 | - | 3 | 403 | |||
BC099374.1 | - | 3 | 412 | |||
CB273987.1 | - | 2 | 520 | |||
Z48496.1 | - | 2 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
