siRNA Details
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021278.2 | NP_067253.1 | 2 | 179 | ||
GenBank mRNA | AK010659.1 | - | 2 | 123 | ||
AK088783.1 | - | 2 | 123 | |||
AK162286.1 | - | 2 | 124 | |||
AK169108.1 | - | 2 | 126 | |||
AK169718.1 | - | 2 | 123 | |||
BC018286.1 | - | 2 | 95 | 3501378 | ||
BQ127748.1 | - | 2 | 179 | |||
X16053.1 | - | 2 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
