siRNA Details
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164745.1 | NP_001158217.1 | 5 | 1077 | ||
NM_008974.4 | NP_033000.1 | 5 | 1108 | |||
GenBank mRNA | AF035644.1 | - | 4 | 559 | ||
AK033092.1 | - | 4 | 813 | |||
AK045093.1 | - | 4 | 810 | |||
AK135662.1 | - | 4 | 314 | |||
AK155895.1 | - | 6 | 1151 | |||
BC086794.1 | - | 4 | 490 | 6811716 | ||
BC087551.1 | - | 6 | 1136 | 6329297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
