siRNA Details
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164745.1 | NP_001158217.1 | 5 | 1082 | ||
NM_008974.4 | NP_033000.1 | 5 | 1113 | |||
GenBank mRNA | AF035644.1 | - | 4 | 564 | ||
AK033092.1 | - | 4 | 818 | |||
AK045093.1 | - | 4 | 815 | |||
AK135662.1 | - | 4 | 319 | |||
AK155895.1 | - | 6 | 1156 | |||
BC086794.1 | - | 4 | 495 | 6811716 | ||
BC087551.1 | - | 6 | 1141 | 6329297 |
