siRNA Details
Gene Symbol: Ptprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029928.2 | NP_084204.2 | 12 | 3107 | ||
GenBank mRNA | AK147439.1 | - | 12 | 3109 | ||
AK147668.1 | - | 12 | 3109 | |||
AY077755.1 | - | 12 | 2985 | |||
BC047086.1 | - | 2 | 167 | |||
BC141006.1 | - | 12 | 2903 | 9055618 | ||
BC145111.1 | - | 12 | 2903 | 9053659 |
