siRNA Details
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011212.3 | NP_035342.3 | 18 | 1963 | ||
GenBank mRNA | AK004602.1 | - | 18 | 1965 | ||
AK132455.1 | - | 17 | 1926 | |||
AK154910.1 | - | 15 | 1515 | |||
D83484.1 | - | 18 | 2300 | |||
U35368.1 | - | 18 | 1963 | |||
U36758.1 | - | 15 | 1512 | |||
U40280.1 | - | 17 | 1703 | |||
U62387.1 | - | 18 | 1865 | |||
Z23053.1 | - | 2 | 95 |
