siRNA Details
Gene Symbol: Ptprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008984.2 | NP_033010.2 | 8 | 1642 | ||
GenBank mRNA | AK129005.1 | - | 6 | 1009 | ||
AK220171.1 | - | 6 | 913 | |||
BC058614.1 | - | 8 | 1652 | 30056619 | ||
BC079621.1 | - | 8 | 1256 | 30536348 | ||
X58287.1 | - | 8 | 1567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
