siRNA Details
Gene Symbol: Ptprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008984.2 | NP_033010.2 | 6 | 1244 | ||
GenBank mRNA | AK129005.1 | - | 4 | 611 | ||
AK220171.1 | - | 4 | 515 | |||
BC058614.1 | - | 6 | 1254 | 30056619 | ||
BC079621.1 | - | 6 | 858 | 30536348 | ||
X58287.1 | - | 6 | 1169 |

More Information
Other Species Targeted

Gene Ontology Categories:
