siRNA Details
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081306.1 | NP_001074775.1 | 12 | 4073 | ||
GenBank mRNA | AJ133130.1 | - | 12 | 3853 | ||
AK141126.1 | - | 1 | 1622 | |||
BC151071.1 | - | 12 | 3798 | 9087984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
