siRNA Details
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008986.2 | NP_033012.1 | 2 | 1123 | ||
GenBank mRNA | AF036249.1 | - | 2 | 1078 | ||
BC110698.1 | - | 2 | 1105 | 30608521 | ||
BC116701.1 | - | 2 | 1065 | 40104877 | ||
BC117527.1 | - | 2 | 1065 | 40104876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
