siRNA Details
Gene Symbol: Pts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011220.2 | NP_035350.1 | 2 | 103 | ||
GenBank mRNA | AF043225.1 | - | 2 | 118 | ||
AK002614.1 | - | 2 | 113 | |||
AK146987.1 | - | 2 | 114 | |||
AK153959.1 | - | 2 | 119 | |||
BC029013.1 | - | 2 | 103 | 5357873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
