siRNA Details
Gene Symbol: Pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008993.2 | NP_033019.2 | 3 | 413 | ||
GenBank mRNA | AF309644.1 | - | 3 | 267 | ||
AK003118.1 | - | 3 | 267 | |||
BC057975.1 | - | 3 | 254 | 5055267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
