siRNA Details
Gene Symbol: IHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002181.3 | NP_002172.2 | 2 | 577 | ||
GenBank mRNA | BC034757.1 | - | 2 | 247 | ||
BC136587.1 | - | 2 | 577 | 9020577 | ||
BC136588.1 | - | 2 | 577 | 9020578 | ||
L38517.1 | - | 1 | 232 |
