siRNA Details
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159725.2 | NP_001153197.2 | 3 | 456 | ||
NM_008998.4 | NP_033024.1 | 3 | 687 | |||
GenBank mRNA | AB232611.1 | - | 2 | 293 | ||
AK009707.1 | - | 3 | 457 | |||
AK075591.1 | - | 3 | 688 | |||
BC013170.1 | - | 3 | 536 | 2648145 | ||
BC051071.1 | - | 3 | 663 | 6307319 | ||
X70804.1 | - | 2 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
