siRNA Details
Gene Symbol: Rab20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011227.1 | NP_035357.1 | 2 | 229 | ||
GenBank mRNA | AB232614.1 | - | 2 | 229 | ||
AK145566.1 | - | 2 | 406 | |||
AK152454.1 | - | 2 | 355 | |||
AK163743.1 | - | 2 | 369 | |||
AK164541.1 | - | 2 | 369 | |||
AK217678.1 | - | 2 | 174 | |||
BC089570.1 | - | 2 | 354 | 6747570 | ||
X80332.1 | - | 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
