siRNA Details
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009003.3 | NP_033029.2 | 4, 5 | 432 | ||
GenBank mRNA | AB232591.1 | - | 4, 5 | 292 | ||
AK136566.1 | - | 4, 5 | 421 | |||
AK145452.1 | - | 4, 5 | 415 | |||
AK161877.1 | - | 4, 5 | 400 | |||
BC138508.1 | - | 4, 5 | 345 | 8861528 | ||
CO806247.1 | - | 448 | ||||
D86563.1 | - | 4, 5 | 406 |

More Information
Other Species Targeted

Gene Ontology Categories:
