siRNA Details
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177411.4 | NP_803130.1 | 3 | 476 | ||
GenBank mRNA | AB232594.1 | - | 2 | 337 | ||
AK081251.1 | - | 3 | 472 | |||
AK083034.1 | - | 3 | 465 | |||
AK144551.1 | - | 3 | 598 | |||
AK197789.1 | - | 1 | 132 | |||
BC137574.1 | - | 2 | 388 | 8860593 | ||
BC137575.1 | - | 2 | 388 | 8860594 | ||
BY727964.1 | - | 3 | 477 | |||
X84239.1 | - | 2 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
