siRNA Details
Gene Symbol: Rab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293652.1 | NP_001280581.1 | 7, 8 | 954 | ||
NM_001293653.1 | NP_001280582.1 | 5, 6 | 615 | |||
NM_001293654.1 | NP_001280583.1 | 6, 7 | 781 | |||
NM_001293655.1 | NP_001280584.1 | 5, 6 | 694 | |||
NM_009005.3 | NP_033031.2 | 5, 6 | 732 | |||
GenBank mRNA | AB232599.1 | - | 4, 5 | 526 | ||
AK005004.1 | - | 5, 6 | 694 | |||
AK057671.1 | - | 5, 6 | 661 | |||
AK135659.1 | - | 6, 7 | 783 | |||
AK139914.1 | - | 5, 6 | 685 | |||
AK148117.1 | - | 5, 6 | 725 | |||
AK151078.1 | - | 5, 6 | 706 | |||
AK152666.1 | - | 7, 8 | 929 | |||
AK154234.1 | - | 5, 6 | 713 | |||
AK158971.1 | - | 5, 6 | 616 | |||
AK159507.1 | - | 5, 6 | 661 | |||
AK159928.1 | - | 5, 6 | 633 | |||
AK178672.1 | - | 2, 3 | 172 | |||
AK183086.1 | - | 2, 3 | 190 | |||
BC004597.1 | - | 5, 6 | 650 | 3584891 | ||
BC086793.1 | - | 5, 6 | 648 | 6414012 | ||
CT010352.1 | - | 4, 5 | 526 | |||
X89650.1 | - | 5, 6 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
