siRNA Details
Gene Symbol: Raet1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009018.1 | NP_033044.1 | 655 | |||
GenBank mRNA | BC132028.1 | - | 617 | 40130305 | ||
BC132032.1 | - | 617 | 40130309 | |||
D64162.1 | - | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
