siRNA Details
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 9 | 769 | ||
NM_033475.3 | NP_258436.2 | 8 | 1114 | |||
GenBank mRNA | AB082927.1 | - | 8 | 1027 | ||
AB232630.1 | - | 8 | 525 | |||
AF322068.1 | - | 8 | 525 | |||
AF327929.1 | - | 9 | 719 | |||
AK160308.1 | - | 1083 | ||||
BC038638.1 | - | 8 | 589 | 5058535 | ||
S72304.1 | - | 6 | 372 |

More Information
Other Species Targeted

Gene Ontology Categories:
