siRNA Details
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009391.3 | NP_033417.1 | 4 | 288 | ||
GenBank mRNA | AF159256.1 | - | 3 | 154 | ||
AK002436.1 | - | 4 | 378 | |||
AK010569.1 | - | 3 | 220 | |||
AK010672.1 | - | 4 | 197 | |||
AK012268.1 | - | 3 | 257 | |||
AK075900.1 | - | 4 | 226 | |||
AK087974.1 | - | 4 | 199 | |||
AK145272.1 | - | 4 | 227 | |||
AK151341.1 | - | 4 | 220 | |||
AK151937.1 | - | 4 | 220 | |||
AK153197.1 | - | 4 | 234 | |||
AK153411.1 | - | 4 | 220 | |||
AK160647.1 | - | 4 | 227 | |||
AK167040.1 | - | 4 | 199 | |||
AK168183.1 | - | 4 | 226 | |||
AK217413.1 | - | 3 | 149 | |||
BC014829.1 | - | 4 | 235 | 3968440 | ||
BC083356.1 | - | 4 | 192 | 6818977 | ||
L32751.1 | - | 3 | 179 | |||
S83456.1 | - | 4 | 265 |

More Information
Other Species Targeted

Gene Ontology Categories:
