siRNA Details
Gene Symbol: Ranbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011239.2 | NP_035369.2 | 4 | 559 | ||
GenBank mRNA | AK002989.1 | - | 4 | 525 | ||
AK008276.1 | - | 4 | 522 | |||
AK088781.1 | - | 4 | 547 | |||
AK151167.1 | - | 4 | 537 | |||
AK153074.1 | - | 4 | 541 | |||
AK166642.1 | - | 4 | 561 | |||
AK167053.1 | - | 4 | 543 | |||
BC061140.1 | - | 4 | 578 | 6775220 | ||
L25255.1 | - | 4 | 432 | |||
X56045.1 | - | 4 | 564 | |||
X56046.1 | - | 4 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
