siRNA Details
Gene Symbol: Rasgrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011242.2 | NP_035372.2 | 14, 15 | 1723 | ||
GenBank mRNA | AK160563.1 | - | 4, 5 | 454 | ||
Y12339.1 | - | 6, 7 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
