siRNA Details
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011249.2 | NP_035379.2 | 19, 20 | 2784 | ||
GenBank mRNA | AK156055.1 | - | 19, 20 | 2763 | ||
BC060124.1 | - | 19, 20 | 2751 | 6408844 | ||
BC069179.1 | - | 11, 12 | 1562 | |||
U27177.1 | - | 19, 20 | 2768 | |||
U33320.1 | - | 19, 20 | 2751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
